3-(2-Hydroxyethyl)benzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(2-Hydroxyethyl)phenyl)boronic acid

OR2100-1G OR2100-250MG
APOSOR2100-1GEA 116 EUR
APOSOR2100-1G APOSOR2100-250MG
3-(2-Hydroxyethyl)benzeneboronic acid 97%
3-(2-Hydroxyethyl)benzeneboronic acid
Formel: C₈H₁₁BO₃
Molecular Weight: 165,98 g/mol
Schmelzpunkt: 90…93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03095123
CAS-Nummer: 647853-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR