3-(7-Methylimidazo[1,2-a]pyridin-2-yl)phenylamine
Lieferant: Apollo Scientific
OR2106-1G
OR2106-250MG
APOSOR2106-1GEA
145
EUR
APOSOR2106-1G
APOSOR2106-250MG
3-(7-Methylimidazo[1,2-a]pyridin-2-yl)phenylamine
3-(7-Methylimidazo[1,2-a]pyridin-2-yl)phenylamine
Formel:
C₁₄H₁₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787116 CAS-Nummer: 54970-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...