2-Brombenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromophenyl)methanol

OR2107-25G OR2107-100G
APOSOR2107-25GEA 21.9 EUR
APOSOR2107-25G APOSOR2107-100G
2-Brombenzylalkohol 98%
2-Brombenzylalkohol
Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004600
CAS-Nummer: 18982-54-2
EINECS: 242-719-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR