3,4-Methylenedioxycinnamic acid

Lieferant: Apollo Scientific
Achtung

OR21080-100G OR21080-25G OR21080-500G
APOSOR21080-100GEA 85.7 EUR
APOSOR21080-100G APOSOR21080-25G APOSOR21080-500G
3,4-Methylenedioxycinnamic acid
3,4-Methylenedioxycinnamic acid
Formel: C₁₀H₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00005837
CAS-Nummer: 2373-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR