5-Methoxypyridin-3-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methoxy-3-pyridinboronsäure
OR2109-1G
OR2109-5G
APOSOR2109-1GEA
26.1
EUR
APOSOR2109-1G
APOSOR2109-5G
5-Methoxypyridin-3-boronsäure
5-Methoxypyridin-3-boronsäure
Formel:
C₆H₈BNO₃ Molecular Weight: 152.95 g/mol Schmelzpunkt: 224…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425977 CAS-Nummer: 850991-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...