8-Methyl-1,2,3,4-tetrahydrochinolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR21112-1G
OR21112-25G
OR21112-5G
APOSOR21112-1GEA
95.7
EUR
APOSOR21112-1G
APOSOR21112-25G
APOSOR21112-5G
8-Methyl-1,2,3,4-tetrahydrochinolin 98%
8-Methyl-1,2,3,4-tetrahydrochinolin
Formel:
C₁₀H₁₃N Molecular Weight: 147,2200 g/mol Siedepunkt: 255…257 °C/717mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00218100 CAS-Nummer: 52601-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...