3,4-Dichlorzimtsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3,4-Dichlorphenyl)propensäure, 3,4-Dichlorcinnamsäure

OR21168-25G OR21168-5G
APOSOR21168-25GEA 29 EUR
APOSOR21168-25G APOSOR21168-5G
3,4-Dichlorzimtsäure
3,4-Dichlorzimtsäure
Formel: C₉H₆Cl₂O₂
Molecular Weight: 217,05 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004385
CAS-Nummer: 1202-39-7
EINECS: 214-866-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR