3-Amino-1-phenyl-1H-pyrazole-4-carbonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR21176-1G
OR21176-250MG
OR21176-5G
OR21176-500MG
APOSOR21176-1GEA
519
EUR
APOSOR21176-1G
APOSOR21176-250MG
APOSOR21176-5G
APOSOR21176-500MG
3-Amino-1-phenyl-1H-pyrazole-4-carbonitrile 95%
3-Amino-1-phenyl-1H-pyrazole-4-carbonitrile
Formel:
C₁₀H₈N₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00828633 CAS-Nummer: 138942-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...