Ethyl-4-amino-2-(methylthio)pyrimidin-5-carboxylat 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-amino-2-(methylthio)pyrimidin-5-carboxylate, Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate

OR2123-25G OR2123-5G
APOSOR2123-25GEA 241 EUR
APOSOR2123-25G APOSOR2123-5G
Ethyl-4-amino-2-(methylthio)pyrimidin-5-carboxylat 97%
Ethyl-4-amino-2-(methylthio)pyrimidin-5-carboxylat
Formel: C₈H₁₁N₃O₂S
Molecular Weight: 213.2568 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090781
CAS-Nummer: 776-53-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR