3,4-Dihydroxy-5-methoxybenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR2136-1G
OR2136-5G
OR2136-25G
APOSOR2136-1GEA
26.1
EUR
APOSOR2136-1G
APOSOR2136-5G
APOSOR2136-25G
3,4-Dihydroxy-5-methoxybenzaldehyd
3,4-Dihydroxy-5-methoxybenzaldehyde
Formel:
C₈H₈O₄ Molecular Weight: 168.1467 g/mol Schmelzpunkt: 131…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016610 CAS-Nummer: 3934-87-0 EINECS: 223-513-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...