4,5-Dihydro-6-methyl-3(2H)-pyridazinone

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methyl-4,5-dihydropyridazin-3(2H)-one

OR21385-25G OR21385-100G
APOSOR21385-25GEA 132 EUR
APOSOR21385-25G APOSOR21385-100G
4,5-Dihydro-6-methyl-3(2H)-pyridazinone
4,5-Dihydro-6-methyl-3(2H)-pyridazinone
Formel: C₅H₈N₂O
Molecular Weight: 112,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006455
CAS-Nummer: 5157-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR