3-Cyano-2-hydroxypyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyano-2-pyridone, 2-Oxo-1,2-dihydropyridine-3-carbonitrile

OR21405-5G OR21405-1G
APOSOR21405-5GEA 352 EUR
APOSOR21405-5G APOSOR21405-1G
3-Cyano-2-hydroxypyridine 98%
3-Cyano-2-hydroxypyridine
Formel: C6H4N2O
Molecular Weight: 120,11 g/mol
Schmelzpunkt: 224…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00661865
CAS-Nummer: 20577-27-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR