4'-Amino-2'-methylacetanilid
Lieferant: Apollo Scientific
|
Achtung
|
OR21431-1G
OR21431-250MG
APOSOR21431-1GEA
49.3
EUR
APOSOR21431-1G
APOSOR21431-250MG
4'-Amino-2'-methylacetanilid
4'-Amino-2'-methylacetanilid
Formel:
C₉H₁₂N₂O Schmelzpunkt: 141…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00276633 CAS-Nummer: 56891-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...