2-Cyano-N-methyl-acetamide

Lieferant: Apollo Scientific
Achtung

OR21454-10G OR21454-1G OR21454-5G
APOSOR21454-10GEA 39.3 EUR
APOSOR21454-10G APOSOR21454-1G APOSOR21454-5G
2-Cyano-N-methyl-acetamide
2-Cyano-N-methyl-acetamide
Formel: C₄H₆N₂O
Molecular Weight: 98.1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090119
CAS-Nummer: 6330-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR