3,4-Dimethyl-o-phenylendiamin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Diamino-3,4-dimethylbenzol, 3,4-Dimethylbenzol-1,2-diamin, 3,4-Dimethyl-1,2-phenylendiamin
OR21475-1G
OR21475-5G
APOSOR21475-1GEA
101
EUR
APOSOR21475-1G
APOSOR21475-5G
3,4-Dimethyl-o-phenylendiamin
3,4-Dimethyl-o-phenylendiamin
Formel:
C₈H₁₂N₂ Molecular Weight: 136,2 g/mol Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051709 CAS-Nummer: 41927-01-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...