4-Brom-2,3-dimethyl-6-nitroanilin
Lieferant: Apollo Scientific
|
Achtung
|
OR21476-25G
OR21476-5G
OR21476-100G
APOSOR21476-25GEA
163
EUR
APOSOR21476-25G
APOSOR21476-5G
APOSOR21476-100G
4-Brom-2,3-dimethyl-6-nitroanilin
4-Brom-2,3-dimethyl-6-nitroanilin
Formel:
C₈H₉BrN₂O₂ Molecular Weight: 245,08 g/mol Schmelzpunkt: 152…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051710 CAS-Nummer: 108485-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...