3-Chlor-6-methoxypyridazin 97+%

Lieferant: Apollo Scientific
Achtung

OR21479-100G
APOSOR21479-100GEA 78.3 EUR
APOSOR21479-100G
3-Chlor-6-methoxypyridazin 97+%
3-Chloro-6-methoxypyridazine
Formel: C₅H₅ClN₂O
Molecular Weight: 144,56 g/mol
Schmelzpunkt: 84…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006467
CAS-Nummer: 1722-10-7
EINECS: 217-019-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR