3-Chlor-6-methylpyridazin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR21484-100G
OR21484-25G
OR21484-5G
APOSOR21484-100GEA
241
EUR
APOSOR21484-100G
APOSOR21484-25G
APOSOR21484-5G
3-Chlor-6-methylpyridazin 98%
3-Chloro-6-methylpyridazine
Formel:
C₅H₅ClN₂ Molecular Weight: 128,56 g/mol Schmelzpunkt: 58…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052905 CAS-Nummer: 1121-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...