6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR21501-25G
OR21501-5G
APOSOR21501-25GEA
348
EUR
APOSOR21501-25G
APOSOR21501-5G
6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
Formel:
C₅H₆N₂O₃ Molecular Weight: 142,12 g/mol Schmelzpunkt: 199…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052030 CAS-Nummer: 27372-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...