1-Methyl-1H-indazol-4-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-1-methyl-1H-indazole
OR2151-1G
OR2151-250MG
APOSOR2151-1GEA
105
EUR
APOSOR2151-1G
APOSOR2151-250MG
1-Methyl-1H-indazol-4-amine
1-Methyl-1H-indazol-4-amine
Formel:
C₈H₉N₃ Molecular Weight: 147.1800 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09870041 CAS-Nummer: 77894-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...