3-Chlor-5-iodbenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR2165-1G
OR2165-5G
OR2165-25G
APOSOR2165-1GEA
21.9
EUR
APOSOR2165-1G
APOSOR2165-5G
APOSOR2165-25G
3-Chlor-5-iodbenzoesäure 98%
3-Chlor-5-iodbenzoesäure
Formel:
C₇H₄ClIO₂ Molecular Weight: 282,46 g/mol Schmelzpunkt: 190…191 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00672966 CAS-Nummer: 289039-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...