4-Bromphenylhydraziniumchlorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(4-Bromphenyl)hydrazin Hydrochlorid, 4-Bromphenylhydrazinhydrochlorid, 4-Bromphenylhydrazin Hydrochlorid

OR21686-100G OR21686-25G OR21686-5G
APOSOR21686-100GEA 82.8 EUR
APOSOR21686-100G APOSOR21686-25G APOSOR21686-5G
4-Bromphenylhydraziniumchlorid
4-Bromphenylhydraziniumchlorid
Formel: C₆H₈BrClN₂
Molecular Weight: 223.50 g/mol
Schmelzpunkt: 220…230 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00012941
CAS-Nummer: 622-88-8
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR