2-(Phenylthio)nicotinsäure

Lieferant: Apollo Scientific
Achtung

OR21698-1G OR21698-25G OR21698-5G
APOSOR21698-1GEA 87 EUR
APOSOR21698-1G APOSOR21698-25G APOSOR21698-5G
2-(Phenylthio)nicotinsäure
2-(Phenylthio)nicotinsäure
Formel: C₁₂H₉NO₂S
Molecular Weight: 231.2704 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052110
CAS-Nummer: 35620-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR