[(8-Chlor-1-naphthyl)sulfanyl]essigsäure

Lieferant: Apollo Scientific

OR21716-1G
APOSOR21716-1GEA 62.5 EUR
APOSOR21716-1G
[(8-Chlor-1-naphthyl)sulfanyl]essigsäure
(8-Chloro-1-naphthylthio)acetic acid
Formel: C₁₂H₉ClO₂S
Molecular Weight: 252.71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043901
CAS-Nummer: 129-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR