2-Cyano-3,3-bismethylsulphanylacrylic acid methyl ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3,3-bis(methylthio)-2-cyanoacrylate

OR21720-10G OR21720-1G OR21720-5G
APOSOR21720-10GEA 426 EUR
APOSOR21720-10G APOSOR21720-1G APOSOR21720-5G
2-Cyano-3,3-bismethylsulphanylacrylic acid methyl ester 95%
2-Cyano-3,3-bismethylsulphanylacrylic acid methyl ester
Formel: C₇H₉NO₂S₂
Molecular Weight: 203,28 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067841
CAS-Nummer: 3490-92-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR