2,4-Dibrombenzylalkohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,4-Dibromophenyl)methanol

OR2174-10G OR2174-1G OR2174-5G
APOSOR2174-10GEA 88.6 EUR
APOSOR2174-10G APOSOR2174-1G APOSOR2174-5G
2,4-Dibrombenzylalkohol
2,4-Dibrombenzylalkohol
Formel: C₇H₆Br₂O
Molecular Weight: 265.98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09996906
CAS-Nummer: 666747-06-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR