Methyl-3-isochinolincarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR21830-1G
OR21830-5G
APOSOR21830-1GEA
55.1
EUR
APOSOR21830-1G
APOSOR21830-5G
Methyl-3-isochinolincarboxylat
Methyl-3-isochinolincarboxylat
Formel:
C₁₁H₉NO₂ Molecular Weight: 187,2000 g/mol Schmelzpunkt: 84…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075138 CAS-Nummer: 27104-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...