2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR21836-1G
OR21836-5G
OR21836-25G
APOSOR21836-1GEA
43.5
EUR
APOSOR21836-1G
APOSOR21836-5G
APOSOR21836-25G
2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid
2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid
Formel:
C₁₂H₁₂O₅ Schmelzpunkt: 124…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677730 CAS-Nummer: 117621-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...