3,5-Dichlorsalicylsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dichlor-2-hydroxybenzoesäure
OR2188-100G
OR2188-25G
APOSOR2188-100GEA
59.6
EUR
APOSOR2188-100G
APOSOR2188-25G
3,5-Dichlorsalicylsäure 99%
3,5-Dichlorsalicylsäure
Formel:
C₇H₄Cl₂O₃ Molecular Weight: 207,01 g/mol Schmelzpunkt: 220…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002442 CAS-Nummer: 320-72-9 EINECS: 206-281-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...