4'-Decylacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Decylphenyl)ethanon, p-Decylacetophenon, 4-Decyl-1-acetylbenzol
OR21901-1G
OR21901-5G
APOSOR21901-1GEA
65.4
EUR
APOSOR21901-1G
APOSOR21901-5G
4'-Decylacetophenon
4'-Decylacetophenon
Formel:
C₁₈H₂₈O Molecular Weight: 260.42 Schmelzpunkt: 33…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173754 CAS-Nummer: 37593-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...