4'-Iodacetanilid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-Iodophenyl)acetamide
OR2193-1G
OR2193-5G
APOSOR2193-1GEA
21.9
EUR
APOSOR2193-1G
APOSOR2193-5G
4'-Iodacetanilid
4'-Iodacetanilid
Formel:
C₈H₈INO Molecular Weight: 261,0597 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016352 CAS-Nummer: 622-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...