3,5-Di-tert-butyl-4-hydroxybenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR21942-100G
OR21942-25G
APOSOR21942-100GEA
59.6
EUR
APOSOR21942-100G
APOSOR21942-25G
3,5-Di-tert-butyl-4-hydroxybenzaldehyd
3,5-Di-tert-butyl-4-hydroxybenzaldehyd
Formel:
C₁₅H₂₂O₂ Molecular Weight: 234,34 g/mol Schmelzpunkt: 187…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008826 CAS-Nummer: 1620-98-0 EINECS: 216-592-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...