3',5'-Di-tert-butyl-4'-hydroxyacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
OR21943-25G
OR21943-5G
APOSOR21943-25GEA
484
EUR
APOSOR21943-25G
APOSOR21943-5G
3',5'-Di-tert-butyl-4'-hydroxyacetophenon
3',5'-Di-tert-butyl-4'-hydroxyacetophenon
Formel:
C₁₆H₂₄O₂ Molecular Weight: 248.37 g/mol Schmelzpunkt: 146…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017520 CAS-Nummer: 14035-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...