3-Brom-4-methoxybenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brom-4-methoxybenzoesäurenitril

OR2200-25G
APOSOR2200-25GEA 78.3 EUR
APOSOR2200-25G
3-Brom-4-methoxybenzonitril 98%
3-Brom-4-methoxybenzonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143097
CAS-Nummer: 117572-79-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR