4-Chlor-6-methoxy-2-methylchinolin
Lieferant: Apollo Scientific
|
Achtung
|
OR22002-1G
OR22002-5G
APOSOR22002-1GEA
252
EUR
APOSOR22002-1G
APOSOR22002-5G
4-Chlor-6-methoxy-2-methylchinolin
4-Chloro-6-methoxy-2-methylquinoline
Formel:
C₁₁H₁₀ClNO Molecular Weight: 207,660 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093863 CAS-Nummer: 50593-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...