5'-Acetamido-2'-hydroxyacetophenon

Lieferant: Apollo Scientific

Synonyme: N-(3-Acetyl-4-hydroxyphenyl)acetamide

OR22019-5G OR22019-10G
APOSOR22019-5GEA 80 EUR
APOSOR22019-5G APOSOR22019-10G
5'-Acetamido-2'-hydroxyacetophenon
5'-Acetamido-2'-hydroxyacetophenon
Formel: C₁₀H₁₁NO₃
Schmelzpunkt: 167…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100490
CAS-Nummer: 7298-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR