3-Hydroxytyrosol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol
OR2203-10G
OR2203-25G
APOSOR2203-10GEA
364
EUR
APOSOR2203-10G
APOSOR2203-25G
3-Hydroxytyrosol 95%
3-Hydroxytyrosol
Formel:
C₈H₁₀O₃ Molecular Weight: 154.17 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD01320529 CAS-Nummer: 10597-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...