3,5-Di-tert-butyl-o-benzochinon 98%

Lieferant: Apollo Scientific
Achtung

OR22087-25G OR22087-5G
APOSOR22087-25GEA 277 EUR
APOSOR22087-25G APOSOR22087-5G
3,5-Di-tert-butyl-o-benzochinon 98%
3,5-Di-tert-butyl-o-benzochinon
Formel: C₁₄H₂₀O₂
Molecular Weight: 220.31 g/mol
Schmelzpunkt: 114…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001647
CAS-Nummer: 3383-21-9
EINECS: 222-189-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR