Diphenylcyclopropenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Diphenylcyclopropen-3-on, 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-on
OR22091-1G
OR22091-5G
OR22091-10G
APOSOR22091-1GEA
21.9
EUR
APOSOR22091-1G
APOSOR22091-5G
APOSOR22091-10G
Diphenylcyclopropenon
Diphenylcyclopropenone
Formel:
C₁₅H₁₀O Molecular Weight: 206,24 g/mol Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001311 CAS-Nummer: 886-38-4 EINECS: 212-948-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...