Methyl-3-formylindole-6-carboxylate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-Formylindole-6-carboxylate

OR2211-1G OR2211-5G OR2211-25G
APOSOR2211-1GEA 69.6 EUR
APOSOR2211-1G APOSOR2211-5G APOSOR2211-25G
Methyl-3-formylindole-6-carboxylate 95+%
Methyl-3-formylindole-6-carboxylate
Formel: C₁₁H₉NO₃
Molecular Weight: 203,197 g/mol
Schmelzpunkt: 220…224 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00216479
CAS-Nummer: 133831-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR