3-Hydroxy-4-iodbenzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

OR22116-1G OR22116-5G
APOSOR22116-1GEA 43.5 EUR
APOSOR22116-1G APOSOR22116-5G
3-Hydroxy-4-iodbenzaldehyd 98%
3-Hydroxy-4-iodobenzaldehyde
Formel: C₇H₅IO₂
Molecular Weight: 248,0179 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00114697
CAS-Nummer: 135242-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR