7-Acetyl-3,4-dihydro-2H-1,5-benzodioxepin

Lieferant: Apollo Scientific
Achtung

OR22118-1G OR22118-10G OR22118-5G
APOSOR22118-1GEA 65.4 EUR
APOSOR22118-1G APOSOR22118-10G APOSOR22118-5G
7-Acetyl-3,4-dihydro-2H-1,5-benzodioxepin
7-Acetyl-3,4-dihydro-2H-1,5-benzodioxepin
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,21 g/mol
Siedepunkt: 120 °C/0,5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00114700
CAS-Nummer: 22776-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR