4-Hydroxy-3-nitrobenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR22139-100G
OR22139-25G
OR22139-500G
APOSOR22139-100GEA
45.1
EUR
APOSOR22139-100G
APOSOR22139-25G
APOSOR22139-500G
4-Hydroxy-3-nitrobenzoesäure 98%
4-Hydroxy-3-nitrobenzoic acid
Formel:
C₇H₅NO₅ Molecular Weight: 183.12 g/mol Schmelzpunkt: 183…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007119 CAS-Nummer: 616-82-0 EINECS: 210-494-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...