3-Brom-2-nitroanilin 98%

Lieferant: Apollo Scientific
Achtung

OR2214-5G OR2214-25G
APOSOR2214-5GEA 29 EUR
APOSOR2214-5G APOSOR2214-25G
3-Brom-2-nitroanilin 98%
3-Brom-2-nitroanilin
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063248
CAS-Nummer: 7138-15-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR