5-Methylpyridazin-3(2H)-one 98%

Lieferant: Apollo Scientific

OR2216-5G
APOSOR2216-5GEA 201 EUR
APOSOR2216-5G
5-Methylpyridazin-3(2H)-one 98%
5-Methylpyridazin-3(2H)-one
Formel: C5H6N2O
Molecular Weight: 110,1139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743644
CAS-Nummer: 54709-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR