2,4-Dimethoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR2224-5G OR2224-25G OR2224-100G
APOSOR2224-5GEA 21.9 EUR
APOSOR2224-5G APOSOR2224-25G APOSOR2224-100G
2,4-Dimethoxyphenylboronsäure
2,4-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181,98 g/mol
Schmelzpunkt: 125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074590
CAS-Nummer: 133730-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR