2-Adamantylamin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Aminotricyclo[3.3.1.1(3,7)]decan Hydrochlorid, 2-Aminoadamantan Hydrochlorid, Tricyclo[3.3.1.1(3,7)]dec-2-ylaminhydrochlorid, 2-Adamantanammoniumchlorid
OR22317-1G
OR22317-25G
OR22317-5G
APOSOR22317-1GEA
26.1
EUR
APOSOR22317-1G
APOSOR22317-25G
APOSOR22317-5G
2-Adamantylamin Hydrochlorid
2-Adamantylamin Hydrochlorid
Formel:
C₁₀H₁₈ClN Molecular Weight: 187,71 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074743 CAS-Nummer: 10523-68-9 EINECS: 234-074-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...