3-Chloro-4-hydroxypropiophenone
Lieferant: Apollo Scientific
Synonyme:
1-(3-chloro-4-hydroxyphenyl)propan-1-one
OR22406-1G
OR22406-250MG
OR22406-500MG
OR22406-2.5G
APOSOR22406-1GEA
432
EUR
APOSOR22406-1G
APOSOR22406-250MG
APOSOR22406-500MG
APOSOR22406-2.5G
3-Chloro-4-hydroxypropiophenone
3-Chloro-4-hydroxypropiophenone
Formel:
C₉H₉ClO₂ Molecular Weight: 184.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00218581 CAS-Nummer: 2892-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...