5-Brom-2-thenoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromthiophen-2-carbonsäure, 5-Brom-2-thiophencarbonsäure
OR22424-100G
OR22424-25G
OR22424-5G
APOSOR22424-100GEA
165
EUR
APOSOR22424-100G
APOSOR22424-25G
APOSOR22424-5G
5-Brom-2-thenoesäure 97%
5-Brom-2-thenoesäure
Formel:
C₅H₃BrO₂S Molecular Weight: 207.05 g/mol Schmelzpunkt: 138…144 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00079725 CAS-Nummer: 7311-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...