tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Tert-butyl 3-(aminomethyl)azetidine-1-carboxylate, 1-BOC-3-(aminomethyl)azetidine, tert-Butyl 3-(aminomethyl)azetidine-1-carboxylate, 3-Aminomethyl-azetidine-1-carboxylic acid tert-butyl ester
OR2248-500MG
OR2248-1G
OR2248-5G
APOSOR2248-500MGEA
21.9
EUR
APOSOR2248-500MG
APOSOR2248-1G
APOSOR2248-5G
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate
tert-Butyl-3-(aminomethyl)azetidine-1-carboxylate
Formel:
C₉H₁₈N₂O₂ Molecular Weight: 186,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861762 CAS-Nummer: 325775-44-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...